Order Entry
Spain
ContactUsLinkComponent
Ethylmagnesium bromide 0.9 M in THF, AcroSeal®
Corrosive
Flammable
Irritant
Health
Ethylmagnesium bromide 0.9 M in THF, AcroSeal®
Certificates
undefined
Corrosive
Flammable
Irritant
Health
Ethylmagnesium bromide 0.9 M in THF, AcroSeal®
Specification Test Results
  • Appearance:
    Brown to grey-brown to brown-black Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    0.85 to 1.15 M