Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, Carbofuran 200 µg/ml in dichloromethane
Pesticide reference standard, Carbofuran 200 µg/ml in dichloromethane
Catalog # PROOCILULM74191.2
CAS Number:  
Pesticide reference standard, Carbofuran 200 µg/ml in dichloromethane
Catalog # PROOCILULM74191.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    1563-66-2
  • Description:
    Carbofuran 200 µg/ml in dichloromethane
  • Formula:
    C₁₂H₁₅NO₃
  • Description:
    Carbofuran

Specifications

About this item