Chlorfenvinphos(2-chloro-1-(2,4-dichlorophenyl)vinyl diethyl phosphate, mixture of cis and trans isomers)
Catalog # EHERC11290000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:359.57 g/mol
EINECS:207-432-0
UN:2810
ADR:6.1,I
CAS Number:470-90-6
Formula:C₁₂H₁₄Cl₃O₄P