Order Entry
Spain
ContactUsLinkComponent
 
Dipentaerythritol 85+%, Technical Grade
 
undefined
Dipentaerythritol 85+%, Technical Grade

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    215 to 225 °C
  • GC:
    ≥85.0 %
  • Hydroxyl value:
    1125 to 1325 mg KOH/g
  • Residue after evaporation:
    ≤0.003 %
  • Substance darkened by H2SO4:
    passes test
  • Titrable acid:
    ≤0.0009 meq/g

 

 

Frequently Bought Together