Order Entry
Spain
ContactUsLinkComponent
Herbicide reference standard, Cyanazine 500 µg/ml in methanol
Herbicide reference standard, Cyanazine 500 µg/ml in methanol
Catalog # PROOSL20234
CAS Number:  
Herbicide reference standard, Cyanazine 500 µg/ml in methanol
Catalog # PROOSL20234
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    21725-46-2
  • Description:
    Cyanazine 500 µg/ml in methanol
  • Formula:
    C₉H₁₃ClN₆
  • Description:
    Cyanazine

Specifications

About this item