Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Entrada de pedido
Estado del pedido
Su cuenta
Cesta
Spain
Entrada de pedido
Estado del pedido
Su cuenta
Cesta
ContactUsLinkComponent
Home
Standards
Organic Standards
Herbicide reference standard, Atrazine 100 µg/ml in n-nonane, CIL
Herbicide reference standard, Atrazine 100 µg/ml in n-nonane, CIL
PROOCILULM72351.2
:
LGC Standards PROMOCHEM
:
Herbicide reference standard, Atrazine 100 µg/ml in n-nonane, CIL
PROOCILULM72351.2
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
1912-24-9
Description:
Atrazine 100 µg/ml in n-nonane
Formula:
C₈H₁₄ClN₅
Description:
Atrazine