Pesticide reference standard, Hexachlorobenzene 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM61301.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:118-74-1
- Description:Hexachlorobenzene 100 µg/ml in n-nonane
- Formula:C₆Cl₆
- Description:Hexachlorobenzene