Order Entry
Spain
ContactUsLinkComponent
 
Irritant
3-Bromo-N-methylaniline ≥97%
Catalog # H55464.06
CAS Number:  
Irritant
3-Bromo-N-methylaniline ≥97%
Catalog # H55464.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
Boiling Pt:253 °C
Density:1.461
Flash Pt:>110 °C (230 °F)
 :186.05 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:66584-32-5
Formula:C₇H₈BrN
MDL Number:MFCD05664376