Order Entry
Spain
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.9%, Ultra Pure Grade
Dimethyl sulphoxide ≥99.9%, Ultra Pure Grade
Catalog # N182-5X10ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.9%, Ultra Pure Grade
Catalog # N182-5X10ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic tube
  • Pk:
    5
  • Seq:
    0005

Specifications

Specification Test Results

  • Acidity Test:
    Pass
  • Congealing Point:
    ≥18.3 °C
  • Appearance:
    Clear colorless liquid
  • Non volatile Residue:
    ≤5 mg
  • Product Lock Test:
    C9481
  • Purity:
    ≥99.9%
  • Refractive Index:
    1.4755 - 1.4775
  • Specific Gravity:
    1.095 - 1.101
  • Water (KF):
    ≤0.1%

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089