Acetic acid 2 mol/l (2 N)
Catalog # REGECH22001
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,III
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152