Order Entry
Spain
ContactUsLinkComponent
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for AAS and ICP
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for AAS and ICP
  CPIIS4400-100081
 :  CPI
 :  
Cadmium standard solution, 1,000 mg/L Cd in dil. nitric acid standard for AAS and ICP
  CPIIS4400-100081
 :  CPI
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd