Order Entry
Spain
ContactUsLinkComponent
Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™
Corrosive
Irritant
Health
Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™
Certificates
undefined
Corrosive
Irritant
Health
Calcium hydroxide 95.0-100.5% (by EDTA titration), powder FCC, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Food GMP Manufactured Product:
    95.0 - 100.5%
  • Meets F.C.C. Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    Passes Test
  • FCC - Assay (Ca(OH)₂) (by EDTA titrn):
    95.0 - 100.5 %
  • Appearance:
    Passes Test
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Carbonate (CO₃):
    Passes Test
  • FCC - Acid-Insoluble Substances:
    ≤0.5 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Lead (Pb):
    ≤2 ppm
  • FCC - Fluoride (F):
    ≤0.005 %
  • FCC - Magnesium and Alkali Salts:
    ≤4.8 %

Specification Test Results

Related Information

Frequently Bought Together