Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Turbidity Standards
Turbidity standards for LAMO meters
Turbidity standards for LAMO meters
Turbidity standards for LAMO meters
Catalog # LAMO1480
Supplier:
LaMotte Chemicals
CAS Number:
Certificates
Expand image
Turbidity standards for LAMO meters
Catalog # LAMO1480
Supplier:
LaMotte Chemicals
CAS Number:
Certificates
Specifications
Pk:
60 mL
Turbidity:
0 NTU/FNU
Description:
Primary turbidity standard (EPA/ISO)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation