Order Entry
Spain
ContactUsLinkComponent
Medicine beakers
Medicine beakers
Catalog # BOET06.103.0101
Supplier:  BOETTGER PAUL
Supplier #: 06.103.0101
undefined
Medicine beakers
Catalog # BOET06.103.0101
Supplier:  BOETTGER PAUL
Supplier #: 06.103.0101
Log in for availability
Select an Option
30 ml
Select an Option
Blue
Select an Option
02
Select an Option
75
About this item
PP, translucent.
  • Highly transparent, optimum visibility of the content
  • Pressure-resistant and unbreakable
  • Snap-on cover with writing area available as an option
Universal beaker for a wide variety of uses. Ideal for use as a measuring beaker for medicine or to collect/distribute tablets.
Frequently Bought Together