Order Entry
Spain
ContactUsLinkComponent
D-Panthenol
D-Panthenol
Catalog # EHERC15844500
Supplier:  EHRENSTORFER
CAS Number:  
D-Panthenol
Catalog # EHERC15844500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :205.25 g/mol
EINECS:201-327-3
CAS Number:81-13-0
Formula:C₉H₁₉NO₄
MDL Number:MFCD00065006