Order Entry
Spain
Orders LinkContactUsLinkComponent
Acetic acid 100%

Acetic acid 100%

Plastic bottle, 10 L
Catalog # PRLS87167.9010
Flammable
Corrosive
Supplier:   PROLABO CMP
Acetic acid 100%
Catalog # PRLS87167.9010
Supplier:  PROLABO CMP
CAS Number:  64-19-7
Acetic acid 100%
Acetic acid 100%

Plastic bottle, 10 L

CAS Number:  64-19-7
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Boiling Pt:118 °C (1013 hPa)
  • Melting Pt:17 °C
  • Density:1.05 g/cm³ (20 °C)
  • Flash Pt:38.5 °C
  • Storage Temperature:Ambient
  • EINECS:200-580-7
  • UN:2789
  • ADR:8,II
  • REACH:01-2119475328-30

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.