Order Entry
Spain
Orders LinkContactUsLinkComponent
Surface protection tiles, BOLA
Surface protection tiles, BOLA
Supplier:   Bohlender
Surface protection tiles, BOLA
Supplier:  Bohlender
Certificates
Surface protection tiles, BOLA
Surface protection tiles, BOLA
8 Options Available Below
Product Details & Documents
For use as a table pad, slideway or for insulation.
  • Protection mats made from PTFE
  • Standard sizes with different thicknesses
  • Temperature resistant from –200 to +250 °C
Product Family Options
Product Information
  • Colour
    L×W
    Thickness
    Availability
    Price
  • White
    300×300 mm
    4,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 103,00 €
    103,00 €
    1 * 1 UN
       103,00 €
  • White
    300×300 mm
    2,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 51,40 €
    51,40 €
    1 * 1 UN
       51,40 €
  • White
    300×300 mm
    5,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 130,00 €
    130,00 €
    1 * 1 UN
       130,00 €
  • White
    300×300 mm
    15,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 312,00 €
    312,00 €
    1 * 1 UN
       312,00 €
  • White
    300×300 mm
    10,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 228,00 €
    228,00 €
    1 * 1 UN
       228,00 €
  • White
    300×300 mm
    8,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 190,00 €
    190,00 €
    1 * 1 UN
       190,00 €
  • White
    300×300 mm
    6,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 154,00 €
    154,00 €
    1 * 1 UN
       154,00 €
  • White
    300×300 mm
    3,00 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 73,80 €
    73,80 €
    1 * 1 UN
       73,80 €