
Methanesulphonic acid 1.5 mol/l in aqueous solution
Catalog # PRLS88758.300
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 L
- Seq:5086
Specifications
About this item
:
:96.11 g/mol
Storage Temperature:Ambient
EINECS:200-898-6
UN:2586
ADR:8,III
:GHS05
CAS Number:75-75-2
Formula:CH₃SO₃H
MDL Number:MFCD00007518