Test strips, potassium, Reflectoquant®, Supelco®
Catalog # 1.16992.0001
Supplier: Merck
CAS Number:
Specifications
- Chemical method:Dipicrylamine
- Pk:50 Tests
- Description:Potassium test
- M-Clarity quality level:MQ100
- Range:250 - 1200 mg/l K
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly