Test strips, calcium, Reflectoquant®, Supelco®
Catalog # 1.16125.0001
Supplier: Merck
CAS Number:
Specifications
- Chemical method:Phthalein complexone
- Pk:50 Tests
- Description:Calcium test
- M-Clarity quality level:MQ100
- Range:5 - 125 mg/l Ca
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly