1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:23978-55-4
:
Formula:C₁₂H₂₆N₂O₄
MDL Number:MFCD00005112
:262.35 g/mol
:M-Clarity™ quality level = MQ200
EINECS:245-965-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Melting Point:112 - 115 °C
- Molar Mass:262.35 g/mol
Specification Test Results
Frequently Bought Together