Ammonium dihydrogen phosphate in aqueous solution mobile phase for HPLC
Catalog # PRLS88619.360
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:115.03 g/mol
EINECS:231-764-5
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396