Order Entry
Spain
ContactUsLinkComponent
Hydrochloric acid 0.125 mol/l (0.125 N)
Hydrochloric acid 0.125 mol/l (0.125 N)
Catalog # VWRS28556.360
Supplier:  VWR Collection
CAS Number:  
Hydrochloric acid 0.125 mol/l (0.125 N)
Catalog # VWRS28556.360
Supplier:  VWR Collection
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324