Zinc oxide ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1314-13-2
:
Formula:ZnO
MDL Number:MFCD00011300
:81.39 g/mol
Merck Index:13,10200
Melting Pt:1975 °C
Density:5.61 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-222-5
ADR:9,III
:M-Clarity™ quality level = MQ300
UN:3077
REACH:01-2119463881-32
:GHS09
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:1975 °C
- Molar Mass:81.37 g/mol
- Bulk Density:200 - 700 kg/m3
- Density:5.61 g/cm3 (20 °C)
- pH:7 (50 g/l, H2O, 20 °C) (slurry)
Specification Test Results
Related Information
Frequently Bought Together