Order Entry
Spain
Orders LinkContactUsLinkComponent
Ammonia solution 5%

Ammonia solution 5%

Glass bottle, 1 L
Catalog # PRLS88572.290
Irritant
Supplier:   PROLABO CMP
Ammonia solution 5%
Catalog # PRLS88572.290
Supplier:  PROLABO CMP
CAS Number:  1336-21-6
Ammonia solution 5%

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.