Ammonia solution 0.08 mol/l (0.08 N)
Catalog # PRLS87114.320
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650