

Divinylbenzene (mixture of isomers) stabilised, Sigma-Aldrich®
: Merck
:1321-74-0
:
:C₁₀H₁₀
:MFCD01778656
:130.19 g/mol
:195 °C (1013 hPa)
:0.91 g/cm3 (20 °C)
:77 °C
:215-325-5
:3082
:9,III
:M-Clarity™ quality level = MQ200
:Stabilised with 4-tert-butylpyrocatechol (TBC)
:GHS07
:GHS09
:Refrigerator
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay isomers (GC, area%):60.0 - 65.0 % (a/a)
- Density (d 20 °C/ 4 °C):0.913 - 0.916
- Assay isomers ethylvinyl benzol (GC, area%):34.0 - 39.0 % (a/a)
- Identity (IR):passes test
- The material is stabilised with 4-tert-Butylpyrocatechol. At time of analytical testing a content of >300 ppm stabiliser was determined.:0.9 hPa (20 °C)
- Flash Point:77 °C
- Explosion Limit:1.1 - 6.2 %(V)
- Density:0.91 g/cm3 (20 °C)
- Ignition Point:470 °C
- Synonyms:DVB
- Hill Formula:C10H10
Frequently Bought Together