
1,4-Bis(hydroxymethyl)cyclohexane (mixture of cis and trans isomers)
Catalog # 8.01610.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:34.6 °C
- Molar Mass:144.22 g/mol
- Boiling Point:286 °C (1013 hPa)
- Vapor Pressure:- 0.24 hPa (20 °C)
- Flash Point:160 °C DIN 51758
- Density:1.04 g/cm3 (20 °C)
- pH:4.2 (250 g/l, H2O, 20 °C)
- Ignition Point:330 °C DIN 51794
- Synonyms:1,4-Cyclohexylenedimethanol, 1,4-Dimethylolcyclohexane, 1,4-Cyclohexanedimethanol
- Hill Formula:C8H16O2
Specification Test Results
About this item
:
:144.21 g/mol
:M-Clarity™ quality level = MQ200
EINECS:203-268-9
:GHS07
Storage Temperature:Ambient
CAS Number:105-08-8
Formula:C₈H₁₆O₂
MDL Number:MFCD00001512