Barium peroxide
: Merck
:1304-29-6
:
:BaO₂
:MFCD00003454
:169.33 g/mol
:12,01018
:450 °C
:4.96 g/cm³ (20 °C)
:Ambient
:215-128-4
:1449
:5.1 (6.1),II
:GHS03
:GHS07
Frequently Bought Together
Sodium sulphate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Zinc chloride ≥97%, TECHNICAL
: VWR Chemicals
Magnesium sulphate, (dried basis) 98.0-102.0% USP
: VWR Chemicals
Ammonium peroxodisulphate (APS) ≥98% ACS
: VWR Chemicals
Calcium chloride dihydrate 97.0-103.0% Ph. Eur.
: VWR Chemicals
Potassium hexacyanoferrate(III) ≥98%
: Thermo Fisher Scientific
Sodium acetate, anhydrous 99%
: Thermo Fisher Scientific