Strontium nitrate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10042-76-9
:
Formula:Sr(NO₃)₂
MDL Number:MFCD00011248
:211.63 g/mol
Merck Index:13,08926
Boiling Pt:645 °C (1013 hPa)
Melting Pt:570 °C
Density:2.113 g/cm³ (20 °C)
UN:1507
ADR:5.1,III
:M-Clarity™ quality level = MQ300
EINECS:233-131-9
:GHS03
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:660 g/l (20 °C)
- Melting Point:570 °C
- Molar Mass:211.63 g/mol
- Bulk Density:1000 kg/m3
- Boiling Point:645 °C
- Density:2.986 g/cm3 (20 °C)
- pH:5 - 7 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Frequently Bought Together