
Sodium tetrachloroaluminate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-16-9
:
Formula:AlCl₄Na
MDL Number:MFCD00050361
:191.78 g/mol
Merck Index:14,8687
Melting Pt:185 °C
Density:2.010
Storage Temperature:Ambient
EINECS:232-050-6
UN:3260
ADR:8,III
:GHS05
Related Information
Frequently Bought Together