Order Entry
Spain
ContactUsLinkComponent
Sodium metabisulphite ≥98.0%, AnalaR NORMAPUR® analytical reagent
Sodium metabisulphite ≥98.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Sodium metabisulphite ≥98.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • As (Arsenic):
    Max. 0.5 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Zn (Zinc):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together