Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7558-80-7
:
Formula:NaH₂PO₄
MDL Number:MFCD00003527
Merck Index:12,08806
Melting Pt:∼650 °C
Density:0.9996 g/cm³ (20 °C)
:119.98 g/mol
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC28013.264 spec_VWRC28013.264 spec_VWRC28013.366 spec_VWRC28013.366
Product specification
Frequently Bought Together