Tubing connectors, elbow-shaped, BOLA
Supplier: Bohlender
Certificates
About this item
Elbow-shaped fitting, made of PTFE, with two connectors for elastic tubing.
- Universal chemical resistance
- Temperature resistance -200 up to 250 °C
- Vacuum: suitable
Frequently Bought Together