Order Entry
Spain
Orders LinkContactUsLinkComponent
Water

Water

Glass bottle, 2,5 L
Catalog # REGERH1020
   Safer Human and Environmental Health
Supplier:   REAGECON
Water
Catalog # REGERH1020
Supplier:  REAGECON
   Safer Human and Environmental Health
CAS Number:  7732-18-5
Water

Glass bottle, 2,5 L

CAS Number:  7732-18-5
Price
43,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    REGERH1020
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.