Order Entry
Spain
ContactUsLinkComponent
Sodium nitrate, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Sodium nitrate, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Supplier:  Merck
Certificates
Sodium nitrate, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    874 g/l (20 °C)
  • Melting Point:
    308 °C
  • Molar Mass:
    84.99 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    2.26 g/cm3 (20 °C)
  • pH:
    5.5 - 8.0 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    5.1
  • Hazard Class Description:
    Oxidizer

Specification Test Results

Related Information

Frequently Bought Together