Sodium nitrate ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7631-99-4
:
Formula:NaNO₃
MDL Number:MFCD00011119
Boiling Pt:380
Melting Pt:308
Density:2.26
Molecular Weight:84.99 g/mol
Storage Temperature:Ambient
EINECS:231-554-3
ADR:5.1 III
Merck Index:14,8647
UN:1498
REACH:01-2119488221-41
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together