Order Entry
Spain
Orders LinkContactUsLinkComponent
Cesium chloride
Glass bottle for solids, 1 kg
  A1098.1000
Cesium chloride
  A1098.1000
 :  7647-17-8
Cesium chloride
Glass bottle for solids, 1 kg
 :  7647-17-8
1 390,00 €
1 * 1 kg
 :  Cesium Chloride
  •  :7647-17-8
  •  :CsCl
  •  :MFCD00010955
  •  :168.36 g/mol
  •  :13,02021
  •  :1303 °C (1013 hPa)
  •  :646 °C
  •  :3.7808 g/cm³ (20 °C)
  •  :Ambient
  •  :231-600-2
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg