Order Entry
Spain
ContactUsLinkComponent
cis-1,2-Dimethylcyclohexane ≥99%
cis-1,2-Dimethylcyclohexane ≥99%
Catalog # 80009442.0001
CAS Number:  
cis-1,2-Dimethylcyclohexane ≥99%
Catalog # 80009442.0001
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :112.22 g/mol
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–90 °C
Density:0.796 g/cm³ (20 °C)
Flash Pt:12 °C (54 °F)
Storage Temperature:Ambient
EINECS:218-621-2
UN:2263
ADR:3,II
CAS Number:2207-01-4
Formula:C₈H₁₆
MDL Number:MFCD00063621