Order Entry
Spain
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,2-Dichlorobenzene-d₄ solution
Supelco® Organic Reference Standard, 1,2-Dichlorobenzene-d₄ solution
Catalog # 48952-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 1,2-Dichlorobenzene-d₄ solution
Catalog # 48952-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml in methanol
  • Pk:
    2.000 µl
  • Standards:
    Certified reference material
  • CAS No.:
    2199-69-1
  • Description:
    1,2-Dichlorobenzene-d₄ solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    151,03
  • Flash point:
    9 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311