Order Entry
Spain
ContactUsLinkComponent
 
Brij® 58 stabilised, Sigma-Aldrich®
 :  Merck
 
Brij® 58 stabilised, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    38 - 42 °C
  • Flash Point:
    149 °C
  • Density:
    1 g/cm3

 

 

Frequently Bought Together