Bis(2-ethylhexyl)adipate ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 103-23-1 Molecular Formula: C22H42O4 Formula Weight: 370.58 Flash Point: 196°(384°F) Refractive Index: 1.4470 Melting Point: -67° Boiling Point: 166-168°/1mm Beilstein Reference: 1803774 MDL No.: MFCD00009496
Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
- CAS Number:103-23-1
- Formula:C₂₂H₄₂O₄
- MDL Number:MFCD00009496
- Molecular Weight:370.57 g/mol
- Boiling Pt:166…168 °C (1 mmHg)
- Melting Pt:–67 °C
- Density:0.990
- Flash Pt:196 °C (384 °F)
- Storage Temperature:Ambient
- EINECS:203-090-1
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 103-23-1 Molecular Formula: C22H42O4 Formula Weight: 370.58 Flash Point: 196°(384°F) Refractive Index: 1.4470 Melting Point: -67° Boiling Point: 166-168°/1mm Beilstein Reference: 1803774 MDL No.: MFCD00009496
Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
- CAS Number:103-23-1
- Formula:C₂₂H₄₂O₄
- MDL Number:MFCD00009496
- Molecular Weight:370.57 g/mol
- Boiling Pt:166…168 °C (1 mmHg)
- Melting Pt:–67 °C
- Density:0.990
- Flash Pt:196 °C (384 °F)
- Storage Temperature:Ambient
- EINECS:203-090-1
Documents
Organic Building Blocks and Intermediates
Application Notes
Product Family Options
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



