Methyl-3-methyl-2-furoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6141-57-7
:
Formula:C₇H₈O₃
MDL Number:MFCD00014110
:140.14 g/mol
Boiling Pt:60 °C (1 mmHg)
Melting Pt:33…37 °C
Flash Pt:82 °C (180 °F)
Storage Temperature:Ambient
EINECS:228-131-0
Related Information
Frequently Bought Together