

Dichloro(methyl)(phenyl)silane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149-74-6
:
Formula:C₇H₈Cl₂Si
MDL Number:MFCD00000490
:191.13 g/mol
Density:1.176 g/cm³
Flash Pt:82 °C
Storage Temperature:Ambient
EINECS:205-746-2
UN:2437
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5180 to 1.5200 (20°C, 589 nm)
- GC:≥97.5 %
- Titration excess HCl:40.0 to 43.5 % (MgO)
- Arsenic (As):≤4 ppm
- Calcium (Ca):≤0.45 %
- Iron (Fe):≤0.02 %
- Acid insoluble substances:≤0.05 %
- Soluble salts:≤1.0 %
Specification Test Results
Frequently Bought Together