Cyclohexylamine for synthesis, Sigma-Aldrich®
: Merck
:108-91-8
:
:C₆H₁₃N
:MFCD00001486
:99.18 g/mol
:13,02758
:134.5 °C (1013 hPa)
:–18 °C
:0.87 g/cm³ (20 °C)
:Ambient
:203-629-0
:2357
:8 (3),II
:M-Clarity™ quality level = MQ200
:27 °C
:GHS02
:GHS05
:GHS08
:GHS07
- Solubility:(20 °C) soluble
- Melting Point:-18 °C
- Molar Mass:99.17 g/mol
- Boiling Point:133 - 134 °C (1013 hPa)
- Vapor Pressure:14 hPa (20 °C)
- Flash Point:27 °C
- Refractive Index:1.458 (20 °C, 589 nm)
- Explosion Limit:1.6 - 9.4 %(V)
- Density:0.87 g/cm3 (20 °C)
- pH:11.5 (100 g/l, H2O, 20 °C)
- Ignition Point:265 °C
- Synonyms:Aminocyclohexane, Cyclohexanamine
- Hill Formula:C6H13N
Frequently Bought Together