Cyclohexylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-91-8
:
Formula:C₆H₁₃N
MDL Number:MFCD00001486
:99.18 g/mol
Merck Index:13,02758
Boiling Pt:133…134 °C (760 mmHg)
Melting Pt:–17 °C
Density:0.8665 g/cm³
Flash Pt:27 °C
EINECS:203-629-0
UN:2357
ADR:8,II
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4580 to 1.4600 (20°C, 589 nm)
- Water:≤0.3 % (K.F.)
- GC:≥98.5 %
Specification Test Results
Related Information
Frequently Bought Together