

m-Ethyltoluene
Catalog # EHERCA13356300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:120.19 g/mol
Boiling Pt:158…160 °C (1013 hPa)
Melting Pt:–95.5 °C
Melting Pt:–95,5 °C
Density:0,869 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-626-8
UN:3295
ADR:3,III
:GHS02
:GHS09
CAS Number:620-14-4
Formula:C₉H₁₂
MDL Number:MFCD00009259