4-tert-Butyltoluene
Catalog # EHERC10931730
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:192.8 °C (1013 hPa)
Melting Pt:–62.5 °C
Density:0,861 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
:148.25 g/mol
EINECS:202-675-9
UN:2667
CAS Number:98-51-1
Formula:C₁₁H₁₆
MDL Number:MFCD00008837
Boiling Pt:192,8 °C (1013 hPa)
Melting Pt:–62,5 °C