Order Entry
Spain
ContactUsLinkComponent
1,3-Diethylbenzene
1,3-Diethylbenzene
Catalog # EHERC12605500
Supplier:  EHRENSTORFER
CAS Number:  
1,3-Diethylbenzene
Catalog # EHERC12605500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :134.22 g/mol
Boiling Pt:181 °C (1013 hPa)
Melting Pt:–84.2 °C
Melting Pt:–84,2 °C
Density:0,8798 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:205-511-4
UN:2049
ADR:3,III
 :GHS02
 :GHS07
CAS Number:141-93-5
Formula:C₁₀H₁₄
MDL Number:MFCD00009260