Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Naphthalene 100 µg/ml in acetonitrile
Organic reference standard, Naphthalene 100 µg/ml in acetonitrile
Catalog # PROONE5055
CAS Number:  
Organic reference standard, Naphthalene 100 µg/ml in acetonitrile
Catalog # PROONE5055
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    91-20-3
  • Description:
    Naphthalene 100 µg/ml in acetonitrile
  • Formula:
    C₁₀H₈
  • Description:
    Naphthalene

Specifications

About this item